Organic reference standard, Ergocalciferol (Vitamin D2, Calciferol) 10 µg/ml in methanol
PROONE9249
:
- Concentration:10 µg/ml in methanol
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:50-14-6
- Description:Ergocalciferol (Vitamin D2, Calciferol)
- Formula:C₂₈H₄₄O
- Description:Ergocalciferol (Vitamin D2, Calciferol)