Order Entry
Spain
ContactUsLinkComponent
Organic reference standard, Musk ketone 10 µg/ml in cyclohexane
Organic reference standard, Musk ketone 10 µg/ml in cyclohexane
Catalog # PROONE0519
CAS Number:  
Organic reference standard, Musk ketone 10 µg/ml in cyclohexane
Catalog # PROONE0519
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    4,5 mL
  • CAS No.:
    81-14-1
  • Description:
    Musk ketone 10 µg/ml in cyclohexane
  • Formula:
    C₁₄H₁₈N₂O₅
  • Description:
    Musk ketone

Specifications

About this item