Alizarine yellow GG (sodium salt) ≥97.0% (by HPLC)
Catalog # TCIAA0578-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 97.0 area%
- Solubility in hot Water:transparency
- Drying loss:max. 10.0 %
- Transition interval (pH):(pale yellow)10.0 to 12.0 (deep yellow)
Specification Test Results
About this item
:
:309.21 g/mol
Merck Index:14,05919
EINECS:209-536-1
CAS Number:584-42-9
Formula:C₁₃H₈N₃NaO₅
MDL Number:MFCD00044746