Order Entry
Spain
ContactUsLinkComponent
Antipain dihydrochloride
Antipain dihydrochloride
Catalog # J63680.X0
CAS Number:  
Antipain dihydrochloride
Catalog # J63680.X0
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mg
  • CAS No.:
    37682-72-7
  • Storage conditions:
    Freezer
  • Form:
    White to pale yellow powder
  • MDL No.:
    MFCD00135957
  • Protein synonyms:
    ([(S)-1-Carboxy-2-phenylethyl]- carbamoyl-Arg-Val-Arg-al)| [(S)-1-Carboxy-2-phenylethyl]- carbamoyl-Arg-Val-Arg-al|[(S)-1-Carboxy-phenethyl]-carbamoyl-Arg-Val-Arg-aldehyde|[(S)-1-Carboxy-2-phenylethyl]carbamoyl-L-arginyl-L-valyl-argininal|L-Valinamide|N-(Nα-Carbonyl-Arg-Val-Arg-al)-Phe
  • Protein/peptide name:
    Antipain dihydrochloride
  • Purity:
    99%
  • Molecular weight:
    677,62 g/mol
  • Molecular formula:
    C₂₇H₄₄N₁₀O₆·₂HCl

Specifications

About this item

A natural protease inhibitor for trypsin, papain and cathepsins A and B.

  • Solubility: 0,01 M in water