1,4-Dibromo-2,5-diethylbenzene ≥98.0% (by GC)
: TCI
:40787-48-2
:
:C₁₀H₁₂Br₂
:MFCD04038416
:292.0103 g/mol
:Refrigerator
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:38.0 to 42.0 deg-C
Frequently Bought Together