

N-Fluorobenzenesulphonimide ≥98.0% (by HPLC, titration analysis) fluorinating reagent
Supplier: TCI
Certificates
About this item
CAS Number:133745-75-2
:
Formula:C₁₂H₁₀FNO₄S₂
MDL Number:MFCD00144885
:315.34 g/mol
:GHS07
:GHS08
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Iodometric Titration):min. 98.0 %
- Melting point:115.0 to 119.0 deg-C
Specification Test Results
Frequently Bought Together