Order Entry
Spain
ContactUsLinkComponent
Blood sedimentation apparatus
Blood sedimentation apparatus
Catalog # HECH40367010
undefined
Blood sedimentation apparatus
Catalog # HECH40367010
Specifications
  • Description:
    Apparatus with 10 Westergren pipettes
  • Pk:
    1
Specifications
About this item
Blood sedimentation apparatus stand of stainless steel with tilting device for rapid sedimentation.
  • Pipettes with fused graduation 'ASSISTENT-PERMANENT'
Ordering information: Complete with blood sedimentation pipettes, blood mixing glasses with rubber stopper, special rubber caps (macro method only) and directions for use.
Packaging: In a box.