Order Entry
Spain
ContactUsLinkComponent
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) ≥98.0% (by GC) free radical
Irritant
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) ≥98.0% (by GC) free radical
Supplier:  TCI
Certificates
undefined
Irritant
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) ≥98.0% (by GC) free radical
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Iodometric Titration):
    min. 97.0 %
  • Melting point:
    69.0 to 75.0 deg-C
  • Solubility in Methanol:
    very faint turbidity

Specification Test Results

Frequently Bought Together