Order Entry
Spain
ContactUsLinkComponent
Dibenzo-24-crown-8-(dibenzo-1,4,7,10,13,16,19,22-octaoxacyclotetracosane) ≥98.0% (by GC)
Dibenzo-24-crown-8-(dibenzo-1,4,7,10,13,16,19,22-octaoxacyclotetracosane) ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
Dibenzo-24-crown-8-(dibenzo-1,4,7,10,13,16,19,22-octaoxacyclotetracosane) ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Solubility in hot Methanol:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together