1,4-Bis(hydroxymethyl)cyclohexane (mixture of cis and trans isomers) cis- and trans-mixture ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:105-08-8
:
Formula:C₈H₁₆O₂
MDL Number:MFCD00001512
:144.21 g/mol
Boiling Pt:162°C
Flash Pt:166°C
EINECS:203-268-9
Specification Test Results
- Appearance:White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
- Purity (GC):min. 99.0 % (total of isomers)
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together