Dichloromethane, anhydrous ≥99.0% (by GC) stabilised
Catalog # TCIAD3478-500ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %
- Specific gravity (20/20):1.3250 to 1.3290
- Refractive index n20/D:1.4220 to 1.4260
- Water:max. 50 ppm
Specification Test Results
About this item
:
:84.93 g/mol
Merck Index:14,06063
Boiling Pt:40°C
Melting Pt:–97°C
Density:1,322 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-838-9
UN:1593
ADR:6.1,III
:Stabilised with 2-methyl-2-butene (iso-amylene)
:GHS07
:GHS08
:GHS09
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881