Order Entry
Spain
ContactUsLinkComponent
 
Irritant
Health
Environment
Dichloromethane ≥99.5% (by GC) stabilised for spectrophotometry, for spectroscopy
Catalog # TCIAD0529-100ML
Supplier:  TCI
CAS Number:  
undefined
Irritant
Health
Environment
Dichloromethane ≥99.5% (by GC) stabilised for spectrophotometry, for spectroscopy
Catalog # TCIAD0529-100ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.5 %
  • Absorbance (240nm):
    max. 0.300
  • Absorbance (250nm):
    max. 0.030
  • Absorbance (270nm):
    max. 0.010

Specification Test Results

About this item

 :
 :84.93 g/mol
Merck Index:14,06063
Boiling Pt:39°C
Melting Pt:–97°C
Density:1,322 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-838-9
UN:1593
ADR:6.1,III
 :Stabilised with 2-methyl-2-butene (iso-amylene)
 :GHS07
 :GHS08
 :GHS09
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881