1,5-Diamino-2-methylpentane ≥98.0% (by GC, titration analysis)
: TCI
: 2-Methyl-1,5-diaminopentane, 2-Methylpentane-1,5-diamine, 1,5-Diamino-2-methylpentane, 2-Methyl-1,5-pentanediamine
:15520-10-2
:
:C₆H₁₆N₂
:MFCD00013460
:78°C
:116.21 g/mol
:85°C
:239-556-6
:2735
:8 III
:GHS05
:GHS07
- Environmentally Preferable:
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Frequently Bought Together