
γ-(N-Morpholino)propanesulphonic acid (MOPS) ≥98.0% (by titrimetric analysis) Good's buffer component for biological research
Supplier: TCI
Certificates
About this item
CAS Number:1132-61-2
:
Formula:C₇H₁₅NO₄S
MDL Number:MFCD00006183
:209.27 g/mol
Merck Index:14,06265
Melting Pt:> 250 °C
Flash Pt: (closed cup)
EINECS:214-478-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Absorbance:max. 0.15 (0.1mol/L, H2O, 260nm)
- Solubility in Water:almost transparency
- Heavy metals (as Pb):max. 5 ppm
Specification Test Results
Frequently Bought Together