Order Entry
Spain
ContactUsLinkComponent
2-Methoxy-1-methylethyl acetate (PGMEA) ≥98.0% (by GC)
2-Methoxy-1-methylethyl acetate (PGMEA) ≥98.0% (by GC)
Catalog # TCIAP1171-500ML
Supplier:  TCI
CAS Number:  
undefined
2-Methoxy-1-methylethyl acetate (PGMEA) ≥98.0% (by GC)
Catalog # TCIAP1171-500ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :132.16 g/mol
Boiling Pt:146°C
Melting Pt:–67 °C
Density:0,968 g/cm³ (20 °C)
Flash Pt:47°C
EINECS:203-603-9
UN:3271
ADR:3,III
 :GHS02
 :GHS07
CAS Number:108-65-6
Formula:C₆H₁₂O₃
MDL Number:MFCD00038500