
L(-)-Tyrosine ≥98.5% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:60-18-4
:
Formula:C₉H₁₁NO₃
MDL Number:MFCD00002606
:181.19 g/mol
Merck Index:14,09839
Boiling Pt:351 °C (1013 hPa)
Melting Pt:285 °C
Density:1,41 g/cm³ (20 °C)
EINECS:200-460-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Optical purity (LC):min. 98.0 ee%
- Purity (Nonaqueous Titration):min. 98.5 %
- Solubility in 1mol/L HCl:almost transparency
Specification Test Results
Frequently Bought Together