Order Entry
Spain
ContactUsLinkComponent
Benzyl butyl phthalate-[ring-D4] 100 µg/ml in n-nonane (98% D), CIL
Benzyl butyl phthalate-[ring-D4] 100 µg/ml in n-nonane (98% D), CIL
Catalog # PROOCILDLM13691.2
CAS Number:  
Benzyl butyl phthalate-[ring-D4] 100 µg/ml in n-nonane (98% D), CIL
Catalog # PROOCILDLM13691.2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    93951-88-3
  • Description:
    Benzyl butyl phthalate-[ring-D4] 100 µg/ml in n-nonane (98% D)
  • Formula:
    C₁₉H₂₀O₄
  • Description:
    Benzyl butyl phthalate-[ring-D4]

Specifications

About this item