3-Ethoxypropionic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4324-38-3
:
Formula:C₅H₁₀O₃
MDL Number:MFCD00058997
EINECS:224-357-9
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together