Order Entry
Spain
ContactUsLinkComponent
Fluoride standard solution, 25 mg/L F- in Water
Fluoride standard solution, 25 mg/L F- in Water
  PRLS85323.360
 :  
Fluoride standard solution, 25 mg/L F- in Water
  PRLS85323.360
 :  

 

  • Concentration:
    25 mg/l
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    5 L
  • Description:
    Fluoride standard solution