Order Entry
Spain
ContactUsLinkComponent
Ethyl-3-phenylglycidate (mixture of cis and trans isomers) cis- and trans-mixture 90%
Irritant
Ethyl-3-phenylglycidate (mixture of cis and trans isomers) cis- and trans-mixture 90%
Certificates
undefined
Irritant
Ethyl-3-phenylglycidate (mixture of cis and trans isomers) cis- and trans-mixture 90%

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5165 to 1.5205 (20°C, 589 nm)
  • GC:
    ≥88 %

Specification Test Results

Related Information

Frequently Bought Together