Order Entry
Spain
ContactUsLinkComponent
2,4,7,9-Tetramethyldec-5-yne-4,7-diol ((±)- and meso mixture) ≥95.0% (by GC)
Irritant
Health
2,4,7,9-Tetramethyldec-5-yne-4,7-diol ((±)- and meso mixture) ≥95.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
Health
2,4,7,9-Tetramethyldec-5-yne-4,7-diol ((±)- and meso mixture) ≥95.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to lump
  • Purity (GC):
    min. 95.0% (total of isomers)

Specification Test Results

Frequently Bought Together