

Chloroform ≥99.8% (by GC, corrected for water content) stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:67-66-3
:
Formula:CHCl₃
MDL Number:MFCD00000826
:119.38 g/mol
Merck Index:13,02160
Boiling Pt:61.7 °C (1013 hPa)
Melting Pt:–63 °C
Density:1.4832 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1888
ADR:6.1,III
:Stabilised with ethanol 1 %
EINECS:200-663-8
REACH:05-2114614906-45
:GHS07
:GHS08
Specification Test Results
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:—
- Assay (CHCl₃) (by GC, exclusive of preservative, corrected for water):≥ 99.8 %
- Acetone, Aldehydes (as (CH₃)₂CO):Passes Test
- Acid and Chloride:Passes Test
- Color (APHA):≤ 10
- Ethanol (CH₃CH₂OH)(by GC):0.7 - 1.0 %
- Free Chlorine:Passes Test
- Lead (Pb):≤ 0.050 ppm
- Residue after Evaporation:≤ 2.0 ppm
- Substances Darkened by H₂SO₄:Passes Test
- Suitability for Use in Dithizone Tests:Passes Test
- Water (by KF, coulometric):≤ 0.01 %
Specification Test Results
Frequently Bought Together