Order Entry
Spain
Orders LinkContactUsLinkComponent
Capillary columns, VF-624ms
Capillary columns, VF-624ms
Supplier:   VARIAN
Capillary columns, VF-624ms
Supplier:  VARIAN
Certificates
Capillary columns, VF-624ms
Capillary columns, VF-624ms
14 Options Available Below
Product Details & Documents
VF-624ms columns set a new standard for the analysis of volatile organic compounds.
  • Improved phase technology reduces bleed, thereby increasing signal-to-noise ratios
  • Especially suited for analysing solvents according to EPA methods 524, 624 and 8260, as well as USP 467
  • Eliminate ghost peaks and unstable baselines for best data accuracy
  • Enhanced selectivity eliminating co-eluters such as benzene and 1,2-dichloroethane for improved productivity
Product Family Options
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • Capillary
    VF-624ms
    0,84 µm
    0,15 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 793,85 €
    793,85 €
    1 * 1 UN
       793,85 €
  • Capillary
    VF-624ms
    0,84 µm
    0,15 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 1 012,77 €
    1 012,77 €
    1 * 1 UN
       1 012,77 €
  • Capillary
    VF-624ms
    1,40 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 1 442,43 €
    1 442,43 €
    1 * 1 UN
       1 442,43 €
  • Capillary
    VF-624ms
    1,80 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 1 411,74 €
    1 411,74 €
    1 * 1 UN
       1 411,74 €
  • Capillary
    VF-624ms
    1,80 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 1 616,34 €
    1 616,34 €
    1 * 1 UN
       1 616,34 €
  • Capillary
    VF-624ms
    3,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 1 616,34 €
    1 616,34 €
    1 * 1 UN
       1 616,34 €
  • Capillary
    VF-624ms
    3,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 1 820,94 €
    1 820,94 €
    1 * 1 UN
       1 820,94 €
  • Capillary
    VF-624ms
    0,84 µm
    0,15 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 1 544,73 €
    1 544,73 €
    1 * 1 UN
       1 544,73 €
  • Capillary
    VF-624ms
    0,84 µm
    0,15 mm
    40 m
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 1 994,85 €
    1 994,85 €
    1 * 1 UN
       1 994,85 €
  • Capillary
    VF-624ms
    1,40 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 2 138,07 €
    2 138,07 €
    1 * 1 UN
       2 138,07 €
  • Capillary
    VF-624ms
    1,40 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 2 383,59 €
    2 383,59 €
    1 * 1 UN
       2 383,59 €
  • Capillary
    VF-624ms
    1,80 µm
    0,32 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 2 373,36 €
    2 373,36 €
    1 * 1 UN
       2 373,36 €
  • Capillary
    VF-624ms
    3,00 µm
    0,53 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 2 475,66 €
    2 475,66 €
    1 * 1 UN
       2 475,66 €
  • Capillary
    VF-624ms
    3,00 µm
    0,53 mm
    75 m
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 2 895,09 €
    2 895,09 €
    1 * 1 UN
       2 895,09 €