Order Entry
Spain
Orders LinkContactUsLinkComponent
Agilent Inlet Septa for Gas Chromatography | Avantor
Inlet Septa for Gas Chromatography
Advanced Green inlet septa eliminate sticking and reduce coring. Bleed temperature optimized (BTO) inlet septa are preconditioned and ready to use with low-bleed, mass spec capillary columns. Our long-life septa are specially formed for extended use, meaning less downtime. Explore Agilent's complete portfolio of gas chromatography septa to ensure your GC is operating at peak performance.
AdvancedInletSepta-400x495.jpg
With our premium non-stick septa, you'll never have to scrape bits of septum out of your inlet.
BleedTemperatureOptimized-400x495.jpg
Bleed temperature optimized septa are specifically designed for operation at temperatures up to 400°C without bleeding into the sample flow path.
GeneralPurposeSepta-400x495.jpg
Agilent’s general-purpose septa are made from an enhanced injection-molded silicone rubber material.
LongLifeSepta-400x495.jpg
With Agilent's premium non-stick septa, you'll never have to scrape bits of septum out of your inlet.
MerlinMicroseal-400x495.jpg
The Merlin Microseal is a microvalve alternative to the conventional silicone rubber septa used in gas chromatography (GC).