Order Entry
Spain
ContactUsLinkComponent
Ferrules.png
Ferrules and Connectors
Ferrules and connectors are critical components to securing a leak-free flow path in your GC. Agilent is proud to offer a wide selection of supplies for your connection needs, including ferrules varying in size and material, unions, column connectors, and our patented self-tightening column nuts.
Columnnut.png
Ferrules and connectors are critical components to securing a leak-free flow path in your GC. Agilent is proud to offer a wide selection of supplies for your connection needs, including ferrules varying in size and material, unions, column connectors, and our patented self-tightening column nuts.
FlexibleMetal.png
Put an end to frustration about GC capillary column connections! With Flexible Metal ferrule, you can confidently make leak free column connections.
GraphiteFerrules.png
Graphite ferrules can withstand temperatures as high as 450°C making them a top choice for high temperature applications.
QuickSwap.png
Agilent QuickSwap capillary flow technology module and preswaged, fused silica tubing restrictors can increase the productivity.
Self_Tightening.png
Spring-driven piston continuously presses against the short graphite/polyimide ferrule, maintaining a leak-free seal even after hundreds of injections
UltimateUnion.png
An Ultimate Union helps to easily connect guard columns or retention gaps to GC columns.
10-press_fit_connectors.png
Press fit column connectors are now treated with Agilent Ultra Inert deactivation to provide a robust and inert flow path.
Vespel-Ferrules.png
Vespel ferrules offer excellent sealing capabilities when making metal or glass connections.