Order Entry
Denmark
ContactUsLinkComponent
Standard Zinc Solution for Atomic Absorption Spectrophotometry (100 times concentrated) Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 100 mL
Catalog # ACSDJPH092
Supplier:  CPACHEM
New Product
Standard Zinc Solution for Atomic Absorption Spectrophotometry (100 times concentrated) Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 100 mL
Catalog # ACSDJPH092
Supplier:  CPACHEM