Order Entry
Denmark
ContactUsLinkComponent
Muffle Furnace L5/14/C550 Tmax1400 with flap door and protective gas connection, over-temperature limiter and thermocouple port in t he rear wall 1 * 1 items
Catalog # 455-3319
Supplier:  NABERTHERM
New Product
Muffle Furnace L5/14/C550 Tmax1400 with flap door and protective gas connection, over-temperature limiter and thermocouple port in t he rear wall 1 * 1 items
Catalog # 455-3319
Supplier:  NABERTHERM