Order Entry
Denmark
ContactUsLinkComponent
Autoclavable Tyvek Breather Bag, With Easy Chevron Peel Feature & Self Seal Closure, With Steam Indicator Dot, Size: 8 x 15 1 * 500 items
Catalog # KACC00815TWBBSSSD
New Product
Autoclavable Tyvek Breather Bag, With Easy Chevron Peel Feature & Self Seal Closure, With Steam Indicator Dot, Size: 8 x 15 1 * 500 items
Catalog # KACC00815TWBBSSSD