Order Entry
Denmark
ContactUsLinkComponent
Ethanolamine CAS:141-43-5 EC:205-483-3 1ml ampoule CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store in a refrigerator at temperatures between 2°C to 8°C 1 * 1 mL
Catalog # ACSDSB8780.1000
Supplier:  CPACHEM
New Product
Ethanolamine CAS:141-43-5 EC:205-483-3 1ml ampoule CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store in a refrigerator at temperatures between 2°C to 8°C 1 * 1 mL
Catalog # ACSDSB8780.1000
Supplier:  CPACHEM