Order Entry
Denmark
ContactUsLinkComponent
Sodium Arsenite 0.05M (0.1N) CRM, ISO 17034 and ISO 17025,,,, Conservation 24 months.,,,, Store under normal laboratory conditions, at temperatures between 15°C to 25°C,,,, 1000 ml bottle 1 * 1 L
Catalog # ACSDZ15201086
Supplier:  CPACHEM
New Product
Sodium Arsenite 0.05M (0.1N) CRM, ISO 17034 and ISO 17025,,,, Conservation 24 months.,,,, Store under normal laboratory conditions, at temperatures between 15°C to 25°C,,,, 1000 ml bottle 1 * 1 L
Catalog # ACSDZ15201086
Supplier:  CPACHEM