Order Entry
Denmark
ContactUsLinkComponent
Human IL-1 beta Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection ant ibody, Streptavidin-HRP, recombinant human IL-1&#0223 ELISA standard, and standard reconstitution buffer. This match 1 * 576 Tests
Catalog # ANTIA270483-696
Supplier:  ANTIBODIES.COM
New Product
Human IL-1 beta Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection ant ibody, Streptavidin-HRP, recombinant human IL-1&#0223 ELISA standard, and standard reconstitution buffer. This match 1 * 576 Tests
Catalog # ANTIA270483-696
Supplier:  ANTIBODIES.COM