Order Entry
Denmark
ContactUsLinkComponent
Rabbit IgG Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-AP, Rabbit IgG ELISA standard, and standard reconstitution buffer. This matched antibody pair kit can 1 * 576 Tests
Catalog # ANTIA270453-696
Supplier:  ANTIBODIES.COM
New Product
Rabbit IgG Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-AP, Rabbit IgG ELISA standard, and standard reconstitution buffer. This matched antibody pair kit can 1 * 576 Tests
Catalog # ANTIA270453-696
Supplier:  ANTIBODIES.COM