Order Entry
Denmark
ContactUsLinkComponent
Rat SFXN1/TCC ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of rat SFXN1/TCC in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA270992-96
Supplier:  ANTIBODIES.COM
New Product
Rat SFXN1/TCC ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of rat SFXN1/TCC in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA270992-96
Supplier:  ANTIBODIES.COM