Order Entry
Denmark
ContactUsLinkComponent
Human Tick-Borne Encephalitis Virus IgG ELISA Kit is an Enzyme-Linked Immunsorbent Assay designed for the in vitro semiquantitative determination of human TBEV IgG in serum and plasma. 1 * 96 Tests
Catalog # ANTIA324342-96
Supplier:  ANTIBODIES.COM
New Product
Human Tick-Borne Encephalitis Virus IgG ELISA Kit is an Enzyme-Linked Immunsorbent Assay designed for the in vitro semiquantitative determination of human TBEV IgG in serum and plasma. 1 * 96 Tests
Catalog # ANTIA324342-96
Supplier:  ANTIBODIES.COM