Order Entry
Denmark
ContactUsLinkComponent
Human Toxoplasma gondii IgG ELISA Kit is an Enzyme-Linked Immunsorbent Assay designed for the in vitro semiquant determination of hu man Toxoplasma gondii IgG in serum and plasma. 1 * 96 Tests
Catalog # ANTIA324345-96
Supplier:  ANTIBODIES.COM
New Product
Human Toxoplasma gondii IgG ELISA Kit is an Enzyme-Linked Immunsorbent Assay designed for the in vitro semiquant determination of hu man Toxoplasma gondii IgG in serum and plasma. 1 * 96 Tests
Catalog # ANTIA324345-96
Supplier:  ANTIBODIES.COM