Order Entry
Denmark
ContactUsLinkComponent
Human Epstein Barr Virus VCA IgM ELISA Kit is an Enzyme-Linked Immunsorbent Assay designed for the in vitro semiquantitative determi nation of human EBV VCA IgM in serum. 1 * 96 Tests
Catalog # ANTIA324357-96
Supplier:  ANTIBODIES.COM
New Product
Human Epstein Barr Virus VCA IgM ELISA Kit is an Enzyme-Linked Immunsorbent Assay designed for the in vitro semiquantitative determi nation of human EBV VCA IgM in serum. 1 * 96 Tests
Catalog # ANTIA324357-96
Supplier:  ANTIBODIES.COM