Order Entry
Denmark
ContactUsLinkComponent
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human NPFF2 Receptor protein to be purified and analysed in vitro. 1 * 10 µG
  ANTIA325254-10
New Product
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human NPFF2 Receptor protein to be purified and analysed in vitro. 1 * 10 µG
  ANTIA325254-10