Order Entry
Denmark
ContactUsLinkComponent
Enfortumab ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of Enfortu mab in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samples. 1 * 96 Tests
Catalog # ANTIA326546-96
Supplier:  ANTIBODIES.COM
New Product
Enfortumab ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of Enfortu mab in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samples. 1 * 96 Tests
Catalog # ANTIA326546-96
Supplier:  ANTIBODIES.COM