Order Entry
Denmark
ContactUsLinkComponent
Human Epstein Barr Virus EA-D IgG IF Test is an Immunofluorescence Assay designed for the in vitro qualititative determination of hu man EA (D) EBV IgG in serum and plasma. 1 * 80 Tests
Catalog # ANTIA324363-80
Supplier:  ANTIBODIES.COM
New Product
Human Epstein Barr Virus EA-D IgG IF Test is an Immunofluorescence Assay designed for the in vitro qualititative determination of hu man EA (D) EBV IgG in serum and plasma. 1 * 80 Tests
Catalog # ANTIA324363-80
Supplier:  ANTIBODIES.COM