Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human GPCR GPR43 protein to be purified and analysed in vitro. 1 * 50 µG
Catalog # ANTIA325176-50
Supplier: ANTIBODIES.COM