Order Entry
Denmark
ContactUsLinkComponent
Human MAG/GMA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human MAG/GMA in tissue homogenates, cell lysates, cell culture supernates or other biological fluids. 1 * 48 Tests
  ANTIA3150-48
New Product
Human MAG/GMA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human MAG/GMA in tissue homogenates, cell lysates, cell culture supernates or other biological fluids. 1 * 48 Tests
  ANTIA3150-48