Order Entry
Denmark
ContactUsLinkComponent
Rat MAG/GMA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat MAG/GMA in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5188-96
Supplier:  ANTIBODIES.COM
New Product
Rat MAG/GMA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat MAG/GMA in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5188-96
Supplier:  ANTIBODIES.COM