Order Entry
Denmark
ContactUsLinkComponent
Monkey Mast Cell Tryptase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determina tion of monkey Mast Cell Tryptase in serum, plasma, and other biological fluids. 1 * 96 Tests
  ANTIA327060-96
New Product
Monkey Mast Cell Tryptase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determina tion of monkey Mast Cell Tryptase in serum, plasma, and other biological fluids. 1 * 96 Tests
  ANTIA327060-96