Order Entry
Denmark
ContactUsLinkComponent
Mouse Mast Cell Tryptase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of mouse Mast Cell Tryptase in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4559-48
Supplier:  ANTIBODIES.COM
New Product
Mouse Mast Cell Tryptase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of mouse Mast Cell Tryptase in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4559-48
Supplier:  ANTIBODIES.COM