Order Entry
Denmark
ContactUsLinkComponent
Chloramphenicol Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Chloramphenic ol in meat. 1 * 48 Tests
Catalog # ANTIA327191-48
Supplier:  ANTIBODIES.COM
New Product
Chloramphenicol Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Chloramphenic ol in meat. 1 * 48 Tests
Catalog # ANTIA327191-48
Supplier:  ANTIBODIES.COM