Order Entry
Denmark
ContactUsLinkComponent
Rat Monoamine Oxidase A/MAO-A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of rat Monoamine Oxidase A/MAO-A in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA8120-48
Supplier:  ANTIBODIES.COM
New Product
Rat Monoamine Oxidase A/MAO-A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of rat Monoamine Oxidase A/MAO-A in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA8120-48
Supplier:  ANTIBODIES.COM