Order Entry
Denmark
ContactUsLinkComponent
Human ACY-1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human ACY-1 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6629-48
Supplier:  ANTIBODIES.COM
New Product
Human ACY-1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human ACY-1 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6629-48
Supplier:  ANTIBODIES.COM