Order Entry
Denmark
Orders LinkContactUsLinkComponent
Thymol blue indicator

Thymol blue indicator

Bottle
Catalog # APOSOR80646-100G
Supplier:   Apollo Scientific
Thymol blue indicator
Catalog # APOSOR80646-100G
Supplier:  Apollo Scientific
CAS Number:  76-61-9
Thymol blue indicator

Bottle

CAS Number:  76-61-9
Price
483,00 kr
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:76-61-9
  • Formula:C₂₇H₃₀O₅S
  • MDL Number:MFCD00005869
  • Molecular Weight:466,59 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.