Order Entry
Denmark
ContactUsLinkComponent
1 component: 3,5-Dichlorophenol [CAS:591-35-5] 25mg/l in Methanol CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store in a refrigerator at temperatures between 2°C to 8°C 1 * 5 Ampoul
Catalog # CMPLRD0117171.5
New Product
1 component: 3,5-Dichlorophenol [CAS:591-35-5] 25mg/l in Methanol CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store in a refrigerator at temperatures between 2°C to 8°C 1 * 5 Ampoul
Catalog # CMPLRD0117171.5