0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
Tork Liquid and Spray Soap Dispenser, Elevation, black, plastic, 112x291x114mm (wxhxd), system S1/S11 1 * 12 items
115-1682
:
Essity
New Product
Tork Liquid and Spray Soap Dispenser, Elevation, black, plastic, 112x291x114mm (wxhxd), system S1/S11 1 * 12 items
115-1682
:
Essity
New Product
Frequently Bought Together
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Wipes for delicate tasks, Kimtech™ Science
: KIMBERLY CLARK
Facial tissues, KLEENEX®
: KIMBERLY CLARK
VWR® pHenomenal® pH Electrodes
: VWR Collection
VWR®, Wipes, Pre-Saturated
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
pH electrodes, InLab® ISM
: Mettler - Toledo
VWR® Spec-Wipe® 3, Cleanroom Wipes, Non Woven
: VWR Collection
pH electrodes, SenTix® PLUS
: WTW
VWR® Spec-Wipe® 3, Cleanroom Wipes, Pre-Saturated
: VWR Collection
Cables
: Mettler - Toledo
Facial tissues, Tork Premium
: Essity
Dispensers for hand towels, Tork
: Essity
Multifunctional air velocity and indoor air quality (IAQ) instrument, testo 440 and Testo 440 differential pressure (dP)
: Testo